Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DXO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014872.2 | NP_001014872.2 | 5 - 6 | 1201 | 90 | |
GenBank mRNA | BC120425.1 | - | 5 - 6 | 1005 | 90 | |
BT020784.1 | - | 5 - 6 | 1111 | 90 | ||
BT020825.1 | - | 5 - 6 | 1182 | 90 | ||
BT021169.1 | - | 5 - 6 | 1189 | 90 | ||
BT021171.1 | - | 5 - 6 | 1206 | 90 | ||
BT021779.1 | - | 5 - 6 | 1201 | 90 | ||
BT021813.1 | - | 4 - 5 | 1274 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |