Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014873.1 | NP_001014873.1 | 3 - 4 | 477 | 65 | |
GenBank mRNA | BC102226.1 | - | 3 - 4 | 440 | 65 | |
BT021509.1 | - | 3 - 4 | 477 | 65 | ||
BT029846.1 | - | 3 - 4 | 384 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |