Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014873.1 | NP_001014873.1 | 4 - 5 | 661 | 77 | |
GenBank mRNA | BC102226.1 | - | 4 - 5 | 624 | 77 | |
BT021509.1 | - | 4 - 5 | 661 | 77 | ||
BT029846.1 | - | 4 - 5 | 568 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |