Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014875.1 | NP_001014875.1 | 8 - 9 | 873 | 95 | |
GenBank mRNA | BC105553.1 | - | 8 - 9 | 910 | 95 | |
BT021021.1 | - | 8 - 9 | 873 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |