Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014887.1 | NP_001014887.1 | 5 - 6 | 543 | 65 | |
GenBank mRNA | BT021777.1 | - | 5 - 6 | 543 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |