Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSME2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014889.1 | NP_001014889.1 | 3 - 4 | 219 | 90 | |
GenBank mRNA | BC102583.1 | - | 3 - 4 | 180 | 90 | |
BT020957.1 | - | 3 - 4 | 219 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |