Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSME2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014889.1 | NP_001014889.1 | 6 - 7 | 426 | 114 | |
GenBank mRNA | BC102583.1 | - | 6 - 7 | 387 | 114 | |
BT020957.1 | - | 6 - 7 | 426 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |