Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPBAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014911.1 | NP_001014911.1 | 5 - 6 | 828 | 77 | |
GenBank mRNA | BT021854.1 | - | 5 - 6 | 828 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |