Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014934.1 | NP_001014934.1 | 1 - 2 | 321 | 131 | |
GenBank mRNA | BC150026.1 | - | 1 - 2 | 291 | 131 | |
BT020790.1 | - | 1 - 2 | 321 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |