Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014949.1 | NP_001014949.1 | 2 - 3 | 604 | 72 | |
GenBank mRNA | BC119880.1 | - | 2 - 3 | 659 | 72 | |
BT020824.1 | - | 2 - 3 | 604 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |