Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014952.1 | NP_001014952.1 | 1 - 2 | 1089 | 64 | |
GenBank mRNA | BT021203.1 | - | 1 - 2 | 1089 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |