Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMAN2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014953.1 | NP_001014953.1 | 2 - 3 | 315 | 74 | |
GenBank mRNA | BC105565.1 | - | 2 - 3 | 314 | 74 | |
BT021149.1 | - | 2 - 3 | 315 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |