Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014957.1 | NP_001014957.1 | 6 - 7 | 725 | 98 | |
GenBank mRNA | BC123510.1 | - | 5 - 6 | 690 | 98 | |
BT021513.1 | - | 6 - 7 | 725 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |