Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTUB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015517.1 | NP_001015517.1 | 3 - 4 | 266 | 69 | |
GenBank mRNA | BT020786.1 | - | 3 - 4 | 266 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |