Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOSC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015529.1 | NP_001015529.1 | 3 - 4 | 230 | 119 | |
GenBank mRNA | BC133440.1 | - | 3 - 4 | 247 | 119 | |
BT020971.1 | - | 3 - 4 | 230 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |