Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015543.2 | NP_001015543.1 | 3 - 4 | 295 | 87 | |
GenBank mRNA | AY911336.1 | - | 3 - 4 | 296 | 87 | |
BC102865.1 | - | 3 - 4 | 299 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |