Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015553.1 | NP_001015553.1 | 1 - 2 | 123 | 60 | |
GenBank mRNA | BC112563.1 | - | 1 - 2 | 109 | 60 | |
BT020871.1 | - | 1 - 2 | 123 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |