Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015572.1 | NP_001015572.1 | 1 - 2 | 35 | 136 | |
GenBank mRNA | BT021074.1 | - | 1 - 2 | 35 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |