Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015585.1 | NP_001015585.1 | 3 - 4 | 390 | 91 | |
GenBank mRNA | BC111248.1 | - | 3 - 4 | 387 | 91 | |
BT021016.1 | - | 3 - 4 | 390 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |