Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015594.3 | NP_001015594.2 | 4 - 5 | 695 | 75 | |
GenBank mRNA | BC103407.1 | - | 4 - 5 | 696 | 75 | |
EV727326.1 | - | 3 - 4 | 663 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |