Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015598.1 | NP_001015598.1 | 2 - 3 | 1361 | 59 | |
GenBank mRNA | BC146253.1 | - | 2 - 3 | 1365 | 59 | |
BT021600.1 | - | 2 - 3 | 1361 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |