Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015607.1 | NP_001015607.1 | 5 - 6 | 964 | 115 | |
GenBank mRNA | BC114866.1 | - | 5 - 6 | 983 | 115 | |
BT020956.1 | - | 5 - 6 | 964 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |