Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015612.1 | NP_001015612.1 | 1 - 2 | 77 | 70 | |
GenBank mRNA | BC102614.1 | - | 1 - 2 | 82 | 70 | |
BT020926.1 | - | 1 - 2 | 77 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |