Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015618.1 | NP_001015618.1 | 2 - 3 | 214 | 75 | |
GenBank mRNA | BC123810.1 | - | 2 - 3 | 217 | 75 | |
BT020763.1 | - | 2 - 3 | 214 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |