Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015619.1 | NP_001015619.1 | 4 - 5 | 777 | 94 | |
GenBank mRNA | BC123799.1 | - | 1 - 2 | 1947 | 94 | |
BC149811.1 | - | 4 - 5 | 772 | 94 | ||
BT020661.1 | - | 4 - 5 | 674 | 94 | ||
BT020700.1 | - | 3 - 4 | 601 | 94 | ||
BT020802.1 | - | 3 - 4 | 568 | 94 | ||
BT020803.1 | - | 2 - 3 | 459 | 94 | ||
BT020853.1 | - | 4 - 5 | 777 | 94 | ||
BT020867.1 | - | 3 - 4 | 567 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |