Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYW3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015620.1 | NP_001015620.1 | 2 - 3 | 299 | 73 | |
GenBank mRNA | BC114025.1 | - | 2 - 3 | 298 | 73 | |
BT020826.1 | - | 2 - 3 | 299 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |