Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015647.1 | NP_001015647.1 | 4 - 5 | 321 | 125 | |
GenBank mRNA | BC102653.1 | - | 4 - 5 | 336 | 125 | |
BT020974.1 | - | 4 - 5 | 321 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |