Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015651.1 | NP_001015651.1 | 2 - 3 | 169 | 94 | |
GenBank mRNA | BC109801.1 | - | 3 - 4 | 196 | 94 | |
BT020960.1 | - | 2 - 3 | 169 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |