Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UFC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015663.1 | NP_001015663.1 | 5 - 6 | 428 | 59 | |
GenBank mRNA | BC103142.1 | - | 5 - 6 | 436 | 59 | |
BT021067.1 | - | 5 - 6 | 428 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |