Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015668.1 | NP_001015668.1 | 7 - 8 | 624 | 140 | |
GenBank mRNA | BC134779.1 | - | 4 - 5 | 230 | 140 | |
BC142022.1 | - | 7 - 8 | 704 | 140 | ||
BT020842.1 | - | 7 - 8 | 624 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |