Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015668.1 | NP_001015668.1 | 9 - 10 | 755 | 85 | |
GenBank mRNA | BC134779.1 | - | 6 - 7 | 361 | 85 | |
BC142022.1 | - | 9 - 10 | 835 | 85 | ||
BT020842.1 | - | 9 - 10 | 755 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |