Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRINP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015669.1 | NP_001015669.1 | 4 - 5 | 948 | 60 | |
GenBank mRNA | BC118187.1 | - | 4 - 5 | 947 | 60 | |
BT020908.1 | - | 4 - 5 | 948 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Inventoried Probe spans exons |