Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRINP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015669.1 | NP_001015669.1 | 5 - 6 | 1062 | 73 | |
GenBank mRNA | BT020908.1 | - | 5 - 6 | 1062 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |