Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCS1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015671.1 | NP_001015671.1 | 7 - 8 | 1318 | 90 | |
GenBank mRNA | BC103135.1 | - | 7 - 8 | 1124 | 90 | |
BT020945.1 | - | 7 - 8 | 1318 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |