Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYOZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015676.1 | NP_001015676.1 | 5 - 6 | 693 | 144 | |
GenBank mRNA | BC103121.1 | - | 5 - 6 | 694 | 144 | |
BT020817.1 | - | 5 - 6 | 693 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |