Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024499.1 | NP_001019670.1 | 5 - 6 | 574 | 90 | |
GenBank mRNA | BC118212.1 | - | 5 - 6 | 568 | 90 | |
BT020772.1 | - | 5 - 6 | 659 | 90 | ||
BT021699.1 | - | 5 - 6 | 574 | 90 | ||
BT030475.1 | - | 2 - 3 | 516 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |