Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024503.1 | NP_001019674.1 | 1 - 2 | 86 | 69 | |
GenBank mRNA | BT021666.1 | - | 1 - 2 | 86 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Bos taurus Probe spans exons |