Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMFG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024537.2 | NP_001019708.1 | 2 - 3 | 145 | 99 | |
GenBank mRNA | AY911328.1 | - | 2 - 3 | 146 | 99 | |
BC103101.1 | - | 2 - 3 | 137 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |