Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VNN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024556.2 | NP_001019727.2 | 2 - 3 | 359 | 89 | |
GenBank mRNA | BC149325.1 | - | 2 - 3 | 360 | 89 | |
BT021888.1 | - | 2 - 3 | 345 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |