Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSRP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024689.3 | NP_001019860.1 | 4 - 5 | 474 | 67 | |
GenBank mRNA | BC103108.1 | - | 4 - 5 | 475 | 67 | |
DQ022067.1 | - | 3 - 4 | 462 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |