Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025314.2 | NP_001020485.1 | 1 - 2 | 112 | 84 | |
GenBank mRNA | AY911339.1 | - | 1 - 2 | 113 | 84 | |
BC102975.1 | - | 1 - 2 | 109 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |