Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025315.1 | NP_001020486.1 | 2 - 3 | 166 | 133 | |
GenBank mRNA | AY911354.1 | - | 2 - 3 | 166 | 133 | |
BC102560.1 | - | 1 | 167 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |