Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TWF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025320.2 | NP_001020491.1 | 2 - 3 | 139 | 147 | |
GenBank mRNA | AY911376.1 | - | 2 - 3 | 140 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |