Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025322.2 | NP_001020493.1 | 3 - 4 | 464 | 66 | |
GenBank mRNA | AY911332.1 | - | 3 - 4 | 465 | 66 | |
BC102096.1 | - | 3 - 4 | 473 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |