Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025339.2 | NP_001020510.1 | 3 - 4 | 312 | 76 | |
GenBank mRNA | AY911383.1 | - | 3 - 4 | 313 | 76 | |
BC103133.1 | - | 3 - 4 | 311 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |