Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025343.1 | NP_001020514.1 | 2 - 3 | 378 | 84 | |
GenBank mRNA | AF502256.1 | - | 2 - 3 | 378 | 84 | |
BC109589.1 | - | 2 - 3 | 297 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |