Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NANOG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025344.1 | NP_001020515.1 | 3 - 4 | 665 | 63 | |
GenBank mRNA | DQ069776.1 | - | 3 - 4 | 665 | 63 | |
DQ126153.1 | - | 2 - 3 | 144 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Bos taurus Probe spans exons |