Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031852.1 | NP_001027022.1 | 5 - 6 | 622 | 94 | |
GenBank mRNA | AY937382.1 | - | 4 - 5 | 481 | 94 | |
AY937383.1 | - | 5 - 6 | 622 | 94 | ||
BC102340.1 | - | 4 - 5 | 524 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |