Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034233.2 | NP_001029405.1 | 6 - 7 | 717 | 69 | |
GenBank mRNA | AY911343.1 | - | 6 - 7 | 720 | 69 | |
BC103328.1 | - | 6 - 7 | 718 | 69 | ||
BC151811.1 | - | 6 - 7 | 735 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |