Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034249.2 | NP_001029421.1 | 8 - 9 | 1169 | 142 | |
GenBank mRNA | BC102926.1 | - | 8 - 9 | 1170 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |